accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOHNN

CAS RN 15309-85-0

IUPAC Name: N-(pyridin-4-yl)benzenesulfonamide
Synonyms: N-(pyridin-4-yl)benzenesulfonamide
Molecular Weight (g/mol): 234.27
Molecular Formula: C11H10N2O2S
InChi Key: SXYATQLMZOZEFK-UHFFFAOYSA-N
SMILES: O=S(=O)(NC1=CC=NC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results