accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1531-77-7

IUPAC Name: 9-thiatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-one
Synonyms: 9-thiatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-one
Molecular Weight (g/mol): 226.29
Molecular Formula: C14H10OS
InChi Key: JGJDEWXZEIHBNW-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2CSC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)