Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,11-Dihydrodibenzo[b,e]thiepin-11-one 98.0+%, TCI America™
SDP

Catalog No. D34165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34165G 5 g
D341625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34165G Supplier TCI America Supplier No. D34165G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1531-77-7
Molecular Formula C14H10OS
Molecular Weight (g/mol) 226.293
MDL Number MFCD02093738
InChI Key JGJDEWXZEIHBNW-UHFFFAOYSA-N
Synonym dibenzo b,e thiepin-11 6h-one, 6,11-dihydrodibenzo b,e thiepin-11-one, dibenz b,e thiepin-11 6h-one, 6h-benzo c 1 benzothiepin-11-one, 1-oxo-6,11-dihydrodibenzo b,e thiepin, dothiepinone, 9-thiatricyclo 9.4.0.0 3 ,? pentadeca-1 15 ,3,5,7,11,13-hexaen-2-one, acmc-1buub
PubChem CID 73741
IUPAC Name 6H-benzo[c][1]benzothiepin-11-one
SMILES C1C2=CC=CC=C2C(=O)C3=CC=CC=C3S1

Specifications

Melting Point 87°C
Color Yellow
Boiling Point 180°C
Quantity 5 g
Formula Weight 226.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6,11-Dihydrodibenzo[b,e]thiepin-11-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.