accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNNNN

CAS RN 15311-09-8

IUPAC Name: 4-[5-(pyridin-4-yl)-1,3,4-thiadiazol-2-yl]pyridine
Synonyms: 4-[5-(pyridin-4-yl)-1,3,4-thiadiazol-2-yl]pyridine
Molecular Weight (g/mol): 240.28
Molecular Formula: C12H8N4S
InChi Key: UQDBUNOBHAOPPJ-UHFFFAOYSA-N
SMILES: S1C(=NN=C1C1=CC=NC=C1)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results