accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 153279-27-7

IUPAC Name: tetrafluorobenzene-1,3-dicarboxamide
Synonyms: tetrafluorobenzene-1,3-dicarboxamide
Molecular Weight (g/mol): 236.13
Molecular Formula: C8H4F4N2O2
InChi Key: JDNPZWSPPAVPPP-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(F)C(C(N)=O)=C(F)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results