CAS RN 153436-53-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tyrphostin AG 1478 98.0+%, TCI America™
Tyrphostin AG 1478 98.0+%, TCI America™, available now.