CAS RN 153436-54-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PD 153035
An extremely potent, cell-permeamble, reversible, ATP-competitive and specific inhibitor of the tyrosine kinase activity of the epidermal growth factor receptor (EGFR; IC50 = 25 pM; K