Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PD 153035

Catalog No. 2344901MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-449-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-449-01MG Supplier MilliporeSigma™ Supplier No. 2344901MG
Add to Cart
Add to Cart

An extremely potent, cell-permeamble, reversible, ATP-competitive and specific inhibitor of the tyrosine kinase activity of the epidermal growth factor receptor (EGFR; IC50 = 25 pM; K

An extremely potent, cell-permeamble, reversible, ATP-competitive and specific inhibitor of the tyrosine kinase activity of the epidermal growth factor receptor (EGFR; IC50 = 25 pM; Ki = 6 pM). Rapidly suppresses autophosphorylation of EGFR at low nanomolar concentrations in fibroblasts or in human epidermoid carcinoma cells. Also selectively blocks EGF-mediated cellular processes including mitogenesis, early gene expression, and oncogenic transformation. A 10mm (500μg/139μl) solution of PD 153035 (Cat. No. 234491) in DMSO is also available.
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 97% by HPLC
Target Proteins EGFR
CAS 153436-54-5
Molecular Formula C16H14BrN3O2
Formula Weight 360.2g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.