accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)(R)CH3(S)HO

CAS RN 15356-60-2

IUPAC Name: (1S,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
Synonyms: (+)-neomenthol
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: NOOLISFMXDJSKH-UTLUCORTSA-N
SMILES: CC(C)[C@@H]1CC[C@@H](C)C[C@@H]1O
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)