Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Menthol, TCI America™
SDP

Catalog No. M0826500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0826500G 500 g
M0826100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0826500G Supplier TCI America Supplier No. M0826500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15356-60-2
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00062983,MFCD00001484
InChI Key NOOLISFMXDJSKH-UTLUCORTSA-N
Synonym +-menthol, d-menthol, 1s,2r,5s-+-menthol, 1s,2r,5s-menthol, racementhol, 1s,2r,5s-2-isopropyl-5-methylcyclohexanol, unii-c6b1oe8p3w, dl-menthol, menthol, +, cyclohexanol, 5-methyl-2-1-methylethyl-, 1s,2r,5s
PubChem CID 165675
ChEBI CHEBI:76306
IUPAC Name (1S,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
SMILES CC(C)[C@@H]1CC[C@@H](C)C[C@@H]1O

Specifications

Melting Point 42°C
Color White
Boiling Point 99°C
Quantity 500 g
Formula Weight 156.27
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Menthol
EINECSNumber (3)-2333
RTECSNumber OT0525000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.