accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSSSSCH3

CAS RN 153561-79-6

IUPAC Name: 5-dodecyl-5'-{5'-dodecyl-[2,2'-bithiophen]-5-yl}-2,2'-bithiophene
Synonyms: 5-dodecyl-5'-{5'-dodecyl-[2,2'-bithiophen]-5-yl}-2,2'-bithiophene
Molecular Weight (g/mol): 667.14
Molecular Formula: C40H58S4
InChi Key: OWLUEDGBXMVFNL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC1=CC=C(S1)C1=CC=C(S1)C1=CC=C(S1)C1=CC=C(CCCCCCCCCCCC)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results