Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5‴-Didodecyl-2,2':5',2″:5″,2‴-quaterthio 96+%, TCI America™
SDP

Catalog No. D4889100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4889100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4889100MG Supplier TCI America Supplier No. D4889100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 153561-79-6
Molecular Formula C40H58S4
Molecular Weight (g/mol) 667.144
InChI Key OWLUEDGBXMVFNL-UHFFFAOYSA-N
PubChem CID 14973124
IUPAC Name 2-dodecyl-5-[5-[5-(5-dodecylthiophen-2-yl)thiophen-2-yl]thiophen-2-yl]thiophene
SMILES CCCCCCCCCCCCC1=CC=C(S1)C2=CC=C(S2)C3=CC=C(S3)C4=CC=C(S4)CCCCCCCCCCCC

Specifications

Melting Point 166°C
Quantity 100 mg
Formula Weight 667.14
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,5′′′-Didodecyl-2,2′:5′,2′′:5′′,2′′
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.