accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFO

CAS RN 1536-23-8

IUPAC Name: (2,3,4,5,6-pentafluorophenyl)(phenyl)methanone
Synonyms: (2,3,4,5,6-pentafluorophenyl)(phenyl)methanone
Molecular Weight (g/mol): 272.17
Molecular Formula: C13H5F5O
InChi Key: HCCPWBWOSASKLG-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(C(=O)C2=CC=CC=C2)C(F)=C1F
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)