Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5,6-Pentafluorobenzophenone 98.0+%, TCI America™
SDP

Catalog No. P09245G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P09245G Supplier TCI America Supplier No. P09245G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1536-23-8
Molecular Formula C13H5F5O
Molecular Weight (g/mol) 272.17
MDL Number MFCD00000294
InChI Key HCCPWBWOSASKLG-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorobenzophenone, pentafluorobenzophenone, perfluorophenyl phenyl methanone, pentafluorophenyl phenyl methanone, methanone, pentafluorophenyl phenyl, benzophenone, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluorophenyl phenyl methanone, 2,3,4,5,6-pentafluorophenyl phenyl ketone, benzoylpentafluorobenzene, acmc-1bvr7
PubChem CID 73755
IUPAC Name (2,3,4,5,6-pentafluorophenyl)(phenyl)methanone
SMILES FC1=C(F)C(F)=C(C(=O)C2=CC=CC=C2)C(F)=C1F

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 137°C
Quantity 5 g
Formula Weight 272.17
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.