accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 153749-89-4

IUPAC Name: tert-butyl 2-cyanopiperidine-1-carboxylate
Synonyms: tert-butyl 2-cyanopiperidine-1-carboxylate
Molecular Weight (g/mol): 210.28
Molecular Formula: C11H18N2O2
InChi Key: LKAJZBMOVZIKHA-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCCC1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results