accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1537876-29-1

IUPAC Name: dipotassium 4,9-bis(4-bromophenyl)-3,5,8,10,13,14-hexaoxa-4,9-diborapentacyclo[7.3.1.1⁴,¹².0²,⁷.0⁶,¹¹]tetradecane-4,9-diuide
Synonyms: dipotassium 4,9-bis(4-bromophenyl)-3,5,8,10,13,14-hexaoxa-4,9-diborapentacyclo[7.3.1.1⁴,¹².0²,⁷.0⁶,¹¹]tetradecane-4,9-diuide
Molecular Weight (g/mol): 585.93
Molecular Formula: C18H14B2Br2K2O6
InChi Key: KFMXNLSRMBTKTI-UHFFFAOYSA-N
SMILES: [K+].[K+].BrC1=CC=C(C=C1)[B-]12OC3C4O[B-]5(OC3C(O1)C(O5)C4O2)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results