Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(4-bromophenylboronic Acid) scyllo-Inositol Complex Dipotassium Tetrahydrate 98.0+%, TCI America™
SDP

Catalog No. B4524200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4524200MG 200 mg
B45241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4524200MG Supplier TCI America Supplier No. B4524200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1537876-29-1
Molecular Formula C18H14B2Br2K2O6
Molecular Weight (g/mol) 585.929
InChI Key KFMXNLSRMBTKTI-UHFFFAOYSA-N
PubChem CID 91972154
SMILES [B-]12(OC3C4C(O1)C5C(C3O[B-](O4)(O5)C6=CC=C(C=C6)Br)O2)C7=CC=C(C=C7)Br.[K+].[K+]

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 585.93
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-bromophenylboronic Acid) scyllo-Inositol Complex Dipotassium Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.