accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3(S)HNFOO

CAS RN 154-08-5

IUPAC Name: (2S)-2-azaniumyl-3-(5-fluoro-1H-indol-3-yl)propanoate
Synonyms: (2S)-2-ammonio-3-(5-fluoro-1H-indol-3-yl)propanoate
Molecular Weight (g/mol): 222.22
Molecular Formula: C11H11FN2O2
InChi Key: INPQIVHQSQUEAJ-VIFPVBQESA-N
SMILES: [NH3+][C@@H](CC1=CNC2=CC=C(F)C=C12)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results