Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-5-Fluorotryptophan, MP Biomedicals

Catalog No. ICN210177 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210177 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210177 Supplier MP Biomedicals, Inc Supplier No. 0521017780
Add to Cart
Add to Cart

Chemical Identifiers

CAS 154-08-5
Molecular Formula C11H11FN2O2
Molecular Weight (g/mol) 222.22
MDL Number MFCD00005649
InChI Key INPQIVHQSQUEAJ-VIFPVBQESA-N
Synonym 5-fluoro-dl-tryptophan, 5-fluorotryptophan, dl-5-fluorotryptophan, 2-amino-3-5-fluoro-1h-indol-3-yl propanoic acid, tryptophan, 5-fluoro, tryptophan, 5-fluoro-, dl, 5-fluoro-d,l-tryptophan, 2-amino-3-5-fluoroindol-3-yl propanoic acid, h-dl-trp 5-f-oh, dl-tryptophan, 5-fluoro
PubChem CID 9577
ChEBI CHEBI:77837
IUPAC Name (2S)-2-azaniumyl-3-(5-fluoro-1H-indol-3-yl)propanoate
SMILES [NH3+][C@@H](CC1=CNC2=CC=C(F)C=C12)C([O-])=O

Specifications

Quantity 100 mg
Formula Weight 222.22
Chemical Name or Material Dl-5-Fluorotryptophan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.