accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 154-17-6

IUPAC Name: (4R,5S,6R)-6-(hydroxymethyl)oxane-2,4,5-triol
Synonyms: (4R,5S,6R)-6-(hydroxymethyl)oxane-2,4,5-triol
Molecular Weight (g/mol): 164.16
Molecular Formula: C6H12O5
InChi Key: PMMURAAUARKVCB-PHUJZJCSNA-N
SMILES: OC[C@H]1OC(O)C[C@@H](O)[C@@H]1O
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (6)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)