CAS RN 1541-23-7
IUPAC Name:
(5E)-hepta-1,5-diene
Synonyms:
(5E)-hepta-1,5-diene
Molecular Weight (g/mol):
96.17
Molecular Formula:
C7H12
InChi Key:
ZGXMNEKDFYUNDQ-GQCTYLIASA-N
SMILES:
C\C=C\CCC=C
1
–
1
of
1
results
1
1,5-Heptadiene (cis- and trans- mixture) 97.0+%, TCI America™
CAS: 1541-23-7 Molecular Formula: C7H12 Molecular Weight (g/mol): 96.17 MDL Number: MFCD00026998 InChI Key: ZGXMNEKDFYUNDQ-GQCTYLIASA-N PubChem CID: 5364394 IUPAC Name: (5E)-hepta-1,5-diene SMILES: C\C=C\CCC=C