Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Heptadiene (cis- and trans- mixture) 97.0+%, TCI America™
SDP

Catalog No. H05515ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05515ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H05515ML Supplier TCI America Supplier No. H05515ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1541-23-7
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
MDL Number MFCD00026998
InChI Key ZGXMNEKDFYUNDQ-GQCTYLIASA-N
PubChem CID 5364394
IUPAC Name (5E)-hepta-1,5-diene
SMILES C\C=C\CCC=C

Specifications

Color Colorless
Quantity 5 mL
UN Number 3295
Formula Weight 96.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,5-Heptadiene (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.