accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOHNO

CAS RN 15414-82-1

IUPAC Name: 5-ethylimidazolidine-2,4-dione
Synonyms: 5-ethylimidazolidine-2,4-dione
Molecular Weight (g/mol): 128.13
Molecular Formula: C5H8N2O2
InChi Key: RSBRXBZGVHQUJK-UHFFFAOYNA-N
SMILES: CCC1NC(=O)NC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results