Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Ethylhydantoin 92.0+%, TCI America™
SDP

Catalog No. E02865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E02865G Supplier TCI America Supplier No. E02865G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15414-82-1
Molecular Formula C5H8N2O2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00059161
InChI Key RSBRXBZGVHQUJK-UHFFFAOYNA-N
PubChem CID 253217
IUPAC Name 5-ethylimidazolidine-2,4-dione
SMILES CCC1NC(=O)NC1=O

Specifications

Melting Point 120°C
Color White
Quantity 5 g
Formula Weight 128.13
Percent Purity ≥92.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Ethylhydantoin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.