CAS RN 154226-60-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ clasto-Lactacystin β-Lactone
A highly specific, cell-permeable, and irreversible 20S proteasome inhibitor.