Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ clasto-Lactacystin β-Lactone

Catalog No. 426102100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-610-2100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-610-2100UG Supplier MilliporeSigma™ Supplier No. 426102100UG
Add to Cart
Add to Cart

A highly specific, cell-permeable, and irreversible 20S proteasome inhibitor.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Target Proteins 20S proteasome
CAS 154226-60-5
Molecular Formula C10H15NO4
Formula Weight 213.2g/mol
Solubility DMSO
Form Solid
Quantity 100 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.