accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OBrN

CAS RN 15430-62-3

IUPAC Name: 2-bromo-2-cyano-N,N-dimethylacetamide
Synonyms: 2-bromo-2-cyano-N,N-dimethylacetamide
Molecular Weight (g/mol): 191.03
Molecular Formula: C5H7BrN2O
InChi Key: HLAJXHYCSPHGGI-UHFFFAOYNA-N
SMILES: CN(C)C(=O)C(Br)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results