Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-2-cyano-N,N-dimethylacetamide, TCI America™
SDP

Catalog No. B10165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B10165G Supplier TCI America Supplier No. B10165G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15430-62-3
Molecular Formula C5H7BrN2O
Molecular Weight (g/mol) 191.028
MDL Number MFCD00040509
InChI Key HLAJXHYCSPHGGI-UHFFFAOYSA-N
Synonym N,N-Dimethyl-2-bromo-2-cyanoacetamide, 2-(N,N-Dimethylcarbamoyl)-2-bromoacetonitrile
PubChem CID 85841
IUPAC Name 2-bromo-2-cyano-N,N-dimethylacetamide
SMILES CN(C)C(=O)C(C#N)Br

Specifications

Melting Point 57°C
Color White-Yellow
Boiling Point 135°C
Quantity 5 g
Formula Weight 191.03
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-2-cyano-N,N-dimethylacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.