accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15435-29-7

IUPAC Name: 2-bromo-6-[(3-bromo-5-chloro-2-hydroxyphenyl)methyl]-4-chlorophenol
Synonyms: bromochlorophen bromochlorophene
Molecular Weight (g/mol): 426.91
Molecular Formula: C13H8Br2Cl2O2
InChi Key: TYBHZVUFOINFDV-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(Cl)C=C1CC1=CC(Cl)=CC(Br)=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results