Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Methylenebis(6-bromo-4-chlorophenol) 95.0+%, TCI America™
SDP

Catalog No. M194025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M194025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M194025G Supplier TCI America Supplier No. M194025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15435-29-7
Molecular Formula C13H8Br2Cl2O2
Molecular Weight (g/mol) 426.91
MDL Number MFCD00152965
InChI Key TYBHZVUFOINFDV-UHFFFAOYSA-N
Synonym Bis(3-bromo-5-chloro-2-hydroxyphenyl)methane, 3,3′C-Dibromo-5,5′C-dichloro-2,2′C-dihydroxydiphenylmethane, Bromochlorophen
PubChem CID 84907
IUPAC Name 2-bromo-6-[(3-bromo-5-chloro-2-hydroxyphenyl)methyl]-4-chlorophenol
SMILES OC1=C(Br)C=C(Cl)C=C1CC1=CC(Cl)=CC(Br)=C1O

Specifications

Melting Point 191°C
Color White-Yellow
Quantity 25 g
Formula Weight 426.91
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Methylenebis(6-bromo-4-chlorophenol)
RTECSNumber SL9710000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.