accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15439-16-4

IUPAC Name: 1,4,8,12-tetraazacyclopentadecane-1,4,8,12-tetraium
Synonyms: 1,4,8,12-tetraazacyclopentadecane-1,4,8,12-tetraium
Molecular Weight (g/mol): 218.39
Molecular Formula: C11H30N4
InChi Key: KUFDRRWNPNXBRF-UHFFFAOYSA-R
SMILES: C1C[NH2+]CCC[NH2+]CC[NH2+]CCC[NH2+]C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results