Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4,8,12-Tetraazacyclopentadecane 97.0+%, TCI America™
SDP

Catalog No. T16915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16915G 5 g
T16911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16915G Supplier TCI America Supplier No. T16915G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15439-16-4
Molecular Formula C11H30N4
Molecular Weight (g/mol) 218.39
MDL Number MFCD00010049
InChI Key KUFDRRWNPNXBRF-UHFFFAOYSA-R
Synonym 1,4,8,12-tetraazacyclopentadecane, cyclal, acmc-1bo39
PubChem CID 472687
IUPAC Name 1,4,8,12-tetraazacyclopentadecane-1,4,8,12-tetraium
SMILES C1C[NH2+]CCC[NH2+]CC[NH2+]CCC[NH2+]C1

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 5 g
Formula Weight 214.36
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,8,12-Tetraazacyclopentadecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.