accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1544-86-1

IUPAC Name: 1-(difluoromethoxy)-4-nitrobenzene
Synonyms: 1-(difluoromethoxy)-4-nitrobenzene
Molecular Weight (g/mol): 189.12
Molecular Formula: C7H5F2NO3
InChi Key: SVGGBARCOQPYMV-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OC(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results