Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Difluoromethoxy)nitrobenzene, TCI America™
SDP

Catalog No. D26015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26015G Supplier TCI America Supplier No. D26015G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1544-86-1
Molecular Formula C7H5F2NO3
Molecular Weight (g/mol) 189.118
MDL Number MFCD00085009
InChI Key SVGGBARCOQPYMV-UHFFFAOYSA-N
Synonym 4-difluoromethoxy nitrobenzene, 1-difluoromethoxy-4-nitrobenzene, 1-difluoromethoxy-4-nitro-benzene, 4-difluoromethoxy-1-nitrobenzene, acmc-209dar, 4-difluoromethoxynitrobenzene, svggbarcoqpymv-uhfffaoysa, 4-nitro-alpha,alpha-difluoroanisole
PubChem CID 2737014
IUPAC Name 1-(difluoromethoxy)-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC(F)F

Specifications

Quantity 5 g
Formula Weight 189.12
Physical Form Crystals
Chemical Name or Material 4-(Difluoromethoxy)nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.