accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15440-98-9

IUPAC Name: 1-(hexyloxy)-4-nitrobenzene
Synonyms: 1-(hexyloxy)-4-nitrobenzene
Molecular Weight (g/mol): 223.27
Molecular Formula: C12H17NO3
InChi Key: VLMYJULPWGTTAV-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results