Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Hexyloxy-4-nitrobenzene 98.0+%, TCI America™
SDP

Catalog No. N037325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N037325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N037325G Supplier TCI America Supplier No. N037325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15440-98-9
Molecular Formula C12H17NO3
Molecular Weight (g/mol) 223.272
MDL Number MFCD00043610
InChI Key VLMYJULPWGTTAV-UHFFFAOYSA-N
Synonym Hexyl 4-Nitrophenyl Ether, 4-Hexyloxynitrobenzene
PubChem CID 84912
IUPAC Name 1-hexoxy-4-nitrobenzene
SMILES CCCCCCOC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Quantity 25 g
Formula Weight 223.27
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1-Hexyloxy-4-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.