accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONNOHNONH3COOCH3

CAS RN 154589-96-5

IUPAC Name: N-{5-methoxy-6-methyl-16,18-dioxo-29-oxa-1,7,17-triazaoctacyclo[12.12.2.1²,⁶.0⁷,²⁸.0⁸,¹³.0¹⁵,¹⁹.0²⁰,²⁷.0²¹,²⁶]nonacosa-8,10,12,14(28),15(19),20(27),21(26),22,24-nonaen-4-yl}-N-methylbenzamide
Synonyms: N-{5-methoxy-6-methyl-16,18-dioxo-29-oxa-1,7,17-triazaoctacyclo[12.12.2.1²,⁶.0⁷,²⁸.0⁸,¹³.0¹⁵,¹⁹.0²⁰,²⁷.0²¹,²⁶]nonacosa-8,10,12,14(28),15(19),20(27),21(26),22,24-nonaen-4-yl}-N-methylbenzamide
Molecular Weight (g/mol): 584.63
Molecular Formula: C35H28N4O5
InChi Key: MQCCJEYZKWZQHU-UHFFFAOYNA-N
SMILES: COC1C(CC2OC1(C)N1C3=CC=CC=C3C3=C1C1=C(C4=C(C=CC=C4)N21)C1=C3C(=O)NC1=O)N(C)C(=O)C1=CC=CC=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results