accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFF(S)OOHNCl

CAS RN 154598-52-4

IUPAC Name: (4S)-6-chloro-4-(2-cyclopropylethynyl)-4-(trifluoromethyl)-2,4-dihydro-1H-3,1-benzoxazin-2-one
Synonyms: efavirenz EFV stocrin C14H9ClF3NO2 sustiva
Molecular Weight (g/mol): 315.68
Molecular Formula: C14H9ClF3NO2
InChi Key: XPOQHMRABVBWPR-ZDUSSCGKSA-N
SMILES: FC(F)(F)[C@]1(OC(=O)NC2=CC=C(Cl)C=C12)C#CC1CC1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results