accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 154607-01-9

IUPAC Name: 4-bromo-2-chlorobenzonitrile
Synonyms: 4-bromo-2-chlorobenzonitrile
Molecular Weight (g/mol): 216.46
Molecular Formula: C7H3BrClN
InChi Key: AYQBMZNSJPVADT-UHFFFAOYSA-N
SMILES: ClC1=C(C=CC(Br)=C1)C#N
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)