CAS RN 1547-36-0
IUPAC Name:
4,4,4-trifluoro-3-hydroxy-3-(trifluoromethyl)butanoic acid
Synonyms:
4,4,4-trifluoro-3-hydroxy-3-(trifluoromethyl)butanoic acid
Molecular Weight (g/mol):
226.07
Molecular Formula:
C5H4F6O3
InChi Key:
NOHJBOWARMTILE-UHFFFAOYSA-N
SMILES:
OC(=O)CC(O)(C(F)(F)F)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid, 97%
4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid, 97%
CAS: 1547-36-0 Molecular Formula: C5H4F6O3 Molecular Weight (g/mol): 226.07 MDL Number: MFCD00040910 InChI Key: NOHJBOWARMTILE-UHFFFAOYSA-N PubChem CID: 15244 IUPAC Name: 4,4,4-trifluoro-3-hydroxy-3-(trifluoromethyl)butanoic acid SMILES: OC(=O)CC(O)(C(F)(F)F)C(F)(F)F