Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid, 97%

Catalog No. AAL0951606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0951606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0951606 Supplier Thermo Scientific Chemicals Supplier No. L0951606
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1547-36-0 | C5H4F6O3 | 226.07 g/mol

4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid is a beta-hydroxy acid undergoes cyclization reaction to prepare beta-lactone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid is a beta-hydroxy acid undergoes cyclization reaction to prepare beta-lactone.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1547-36-0
Molecular Formula C5H4F6O3
Molecular Weight (g/mol) 226.07
MDL Number MFCD00040910
InChI Key NOHJBOWARMTILE-UHFFFAOYSA-N
PubChem CID 15244
IUPAC Name 4,4,4-trifluoro-3-hydroxy-3-(trifluoromethyl)butanoic acid
SMILES OC(=O)CC(O)(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 85°C to 89°C
Quantity 5 g
UN Number UN3261
Beilstein 1911706
Synonym 3,3-bis trifluoromethyl-3-hydroxypropionic acid, 4,4,4-trifluoro-3-hydroxy-3-trifluoromethyl butanoic acid, butyric acid, 3-hydroxy-4,4,4-trifluoro-3-trifluoromethyl, 3-hydroxy-4,4,4-trifluoro-3-trifluoromethyl butyric acid, 4,4,4-trifluoro-3-hydroxy-3-trifluoromethyl butyric acid, 4,4,4-trifluoro-3-hydroxy-3-trifluoromethylbutyric acid, butanoic acid, 4,4,4-trifluoro-3-hydroxy-3-trifluoromethyl, butanoic acid,4,4,4-trifluoro-3-hydroxy-3-trifluoromethyl, acmc-20anh2, 3,3-bis trifluoromethyl-3-hydroxypropanoic acid
Solubility Information Soluble in water.
Formula Weight 226.08
Percent Purity 97%
Chemical Name or Material 4,4,4-Trifluoro-3-hydroxy-3-(trifluoromethyl)butyric acid
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECSNumber ET4800000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.