accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15471-17-7

IUPAC Name: 1-(3-sulfonatopropyl)pyridin-1-ium
Synonyms: 1-(3-sulfonatopropyl)pyridin-1-ium
Molecular Weight (g/mol): 201.24
Molecular Formula: C8H11NO3S
InChi Key: REEBJQTUIJTGAL-UHFFFAOYSA-N
SMILES: [O-]S(=O)(=O)CCC[N+]1=CC=CC=C1
Percent Purity 
  • (2)
  • (4)
  • (2)
  • (2)
Quantity 
  • (3)
  • (4)
  • (1)
  • (2)
Grade 
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (4)
  • (4)