Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(1-Pyridinio)-1-propanesulfonate

Catalog No. 8280450G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
82-804-50G 50 g
82-804-250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 82-804-50G Supplier Sigma Aldrich Supplier No. 8280450G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-(1-pyridinio)-1-propanesulfonate can be used:
  • As a bronsted acid ionic liquid catalyst in combination with h2so4 and h3po4 for the dehydration of glycerol to acrolein via semi-batch reaction technique
  • As a catalyst in the form of sulfonated heteropoly acid salts for the synthesis of isoamyl isovalerate
  • In the preparation of molybdenum imido alkylidene nhc catalysts, which are used in biphasic olefin metathesis
  • To prepare water-stable so3h-functionalized br++nsted acidic task-specific ionic liquids (tsils)

Specifications

CAS 15471-17-7
Molecular Formula C8H11NO3S
Linear Formula C8H11NO3S
MDL Number MFCD00064468
Quantity 50 g
Synonym 1-(3-Sulfopropyl)pyridinium betain; NDSB 201; PPS
Molecular Weight (g/mol) 201.24
Percent Purity ≥97.0% (N)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.