accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15486-96-1

IUPAC Name: 3-bromopropanoyl chloride
Synonyms: 3-bromopropanoyl chloride
Molecular Weight (g/mol): 171.42
Molecular Formula: C3H4BrClO
InChi Key: IHBVNSPHKMCPST-UHFFFAOYSA-N
SMILES: ClC(=O)CCBr
Quantity 
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)