Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Bromopropionyl chloride

Catalog No. 14251425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-251-425G 25 g
14-251-4100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-251-425G Supplier Sigma Aldrich Supplier No. 14251425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-bromopropionyl chloride reacts with poly(ethylene glycol) methacrylate to yield brominated poly(ethylene glycol) methacrylate
  • It causes the acylation of 4-aryl-substituted 3,4-di-hydropyrimidine(1h)-2-thiones to give bicyclic pyrimido[2,1-b][1,3]thiazines.3-bromopropionyl chloride was used in the preparation of 1-chloro-4-bromo-2-butanone

Specifications

CAS 15486-96-1
Density 1.701 g/mL (at 25°C (literature))
Boiling Point 55°C to 57°C (17 mmHg)
Molecular Formula C3H4BrClO
Refractive Index n20/D 1.49 (literature)
Linear Formula BrCH2CH2COCl
MDL Number MFCD00000746
Quantity 25 g
Molecular Weight (g/mol) 171.42
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.