accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHOOHSOOH(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 154978-38-8

IUPAC Name: 4-[(1-carboxy-4-hydroxynonadeca-6,8,10,13-tetraen-5-yl)sulfanyl]benzoic acid
Synonyms: 4-[(1-carboxy-4-hydroxynonadeca-6,8,10,13-tetraen-5-yl)sulfanyl]benzoic acid
Molecular Weight (g/mol): 472.64
Molecular Formula: C27H36O5S
InChi Key: PKJINWOACFYDQN-UHFFFAOYNA-N
SMILES: CCCCCC=CCC=CC=CC=CC(SC1=CC=C(C=C1)C(O)=O)C(O)CCCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results