Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAY-u 9773, R&D Systems™
SDP

Catalog No. 313850U
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-385-0U 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 31-385-0U Supplier R&D Systems Supplier No. 3138/50U

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Dual CysLT1 and CysLT2 antagonist

Cysteinyl leukotriene receptor antagonist with equal affinity at both CysLT 1 and CysLT 2 receptors (pK B values are 6.8 and 6.5 respectively). Competitive antagonist of LTC 4 - and LTE 4 -induced contractions of the guinea pig trachea.

Chemical Identifiers

CAS 154978-38-8
Molecular Formula C27H36O5S
Molecular Weight (g/mol) 472.64
InChI Key PKJINWOACFYDQN-DYXAOADDSA-N
Synonym 6 r-4-carboxyphenylthio-5 s-hydroxy-7 e ,9 e ,11 e ,14 z-eicosatetraenoic acid
PubChem CID 71312218
IUPAC Name 4-[(4S,5R,6E,8E,10E,13E)-1-carboxy-4-hydroxynonadeca-6,8,10,13-tetraen-5-yl]sulfanylbenzoic acid
SMILES CCCCCC=CCC=CC=CC=CC(C(CCCC(=O)O)O)SC1=CC=C(C=C1)C(=O)O

Specifications

Quantity 50 μg
Formula Weight 472.64
Percent Purity >98%
Product Line Tocris
Chemical Name or Material BAY-u 9773
Recommended Storage Store at -80°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.