accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15499-84-0

IUPAC Name: 4-[9-(4-aminophenyl)-9H-fluoren-9-yl]aniline
Synonyms: 4-[9-(4-aminophenyl)fluoren-9-yl]aniline
Molecular Weight (g/mol): 348.45
Molecular Formula: C25H20N2
InChi Key: KIFDSGGWDIVQGN-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)