Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Bis(4-aminophenyl)fluorene (purified by sublimation) 99.0+%, TCI America™
SDP

Catalog No. B26541G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B26541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B26541G Supplier TCI America Supplier No. B26541G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15499-84-0
Molecular Formula C25H20N2
Molecular Weight (g/mol) 348.449
MDL Number MFCD00039156
InChI Key KIFDSGGWDIVQGN-UHFFFAOYSA-N
Synonym 4,4′C-(9-Fluorenylidene)dianiline
PubChem CID 631552
IUPAC Name 4-[9-(4-aminophenyl)fluoren-9-yl]aniline
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2(C4=CC=C(C=C4)N)C5=CC=C(C=C5)N

Specifications

Melting Point 237°C
Color White
Quantity 1 g
Formula Weight 348.45
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(4-aminophenyl)fluorene (purified by sublimation)
EINECSNumber (4)-1649
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.