CAS RN 1550-44-3
IUPAC Name:
2,2-difluoroethyl acetate
Synonyms:
2,2-difluoroethyl acetate
Molecular Weight (g/mol):
124.09
Molecular Formula:
C4H6F2O2
InChi Key:
PFJLHSIZFYNAHH-UHFFFAOYSA-N
SMILES:
CC(=O)OCC(F)F
1
–
1
of
1
results
1
2,2-Difluoroethyl Acetate 98.0+%, TCI America™
CAS: 1550-44-3 Molecular Formula: C4H6F2O2 Molecular Weight (g/mol): 124.087 MDL Number: MFCD00069133 InChI Key: PFJLHSIZFYNAHH-UHFFFAOYSA-N Synonym: Acetatic Acid 2,2-Difluoroethyl Ester PubChem CID: 137074 IUPAC Name: 2,2-difluoroethyl acetate SMILES: CC(=O)OCC(F)F